Anti-CECR6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: Biorbyt
BIRBORB5408-100EA 0 EUR
BIRBORB5408-100
Anti-CECR6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: CECR6 (cat eye syndrome chromosome region, candidate 6)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human