Anti-SAMD3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB5420-100EA 385 EUR
BIRBORB5420-100
Anti-SAMD3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: SAMD3 (sterile alpha motif domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR