Anti-PPARG Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB5574-100EA 0 EUR
BIRBORB5574-100
Anti-PPARG Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:2000

Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR