Anti-CDKN1C Rabbit polyclonal antibody
Toimittaja: Biorbyt
BIRBORB5837-100EA 0 EUR
BIRBORB5837-100
Anti-CDKN1C Rabbit polyclonal antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CDKN1C (cyclin-dependent kinase inhibitor 1C (p57, Kip2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat