Anti-PCK2 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB5876-100EA 0 EUR
BIRBORB5876-100 BIRBORB5876-200
Anti-PCK2 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: PCK2 (phosphoenolpyruvate carboxykinase 2 (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat