Anti-CYP11B1 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB5937-200EA 0 EUR
BIRBORB5937-200 BIRBORB5937-100
Anti-CYP11B1 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CYP11B1 (cytochrome P450, family 11, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now