Anti-cTnT Rabbit polyclonal antibody
Toimittaja: Biorbyt
BIRBORB6180-100EA 0 EUR
BIRBORB6180-100 BIRBORB6180-200
Anti-cTnT Rabbit polyclonal antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: cTnT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now