Anti-OLR1 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB6324-200EA 0 EUR
BIRBORB6324-200 BIRBORB6324-100
Anti-OLR1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: OLR1 (oxidized low density lipoprotein (lectin-like) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now