Anti-SYK Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB7034-100EA 0 EUR
BIRBORB7034-100
Anti-SYK Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR