Anti-NRXN2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: Biorbyt
BIRBORB7954-100EA 0 EUR
BIRBORB7954-100
Anti-NRXN2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: NRXN2 (neurexin 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now