Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: Biorbyt
BIRBORB8121-100EA 0 EUR
BIRBORB8121-100
Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR