Anti-NCAM1 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB8197-100EA 0 EUR
BIRBORB8197-100
Anti-NCAM1 Mouse Monoclonal Antibody
Vasta-aineet
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR