Anti-APOA1 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB8245-100EA 0 EUR
BIRBORB8245-100
Anti-APOA1 Mouse Monoclonal Antibody
Vasta-aineet
Western Blot: 1:10001:3000

Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Murine)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR