Anti-MDA-LDL Mouse Monoclonal Antibody [clone: 3G5]

Toimittaja: Biorbyt
BIRBORB8258-100EA 0 EUR
BIRBORB8258-100
Anti-MDA-LDL Mouse Monoclonal Antibody [clone: 3G5]
Vasta-aineet
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: MDA-LDL
Clonality: monoclonal
Clone: 3G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Murine)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR