Anti-APOA4 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB8556-100EA 0 EUR
BIRBORB8556-100
Anti-APOA4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: APOA4 (apolipoprotein A-IV)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR