Anti-PML Rabbit Polyclonal Antibody (FITC (Fluorescein))
Toimittaja: Biorbyt
BIRBORB9613-100EA 0 EUR
BIRBORB9613-100
Anti-PML Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: PML (promyelocytic leukemia)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat