Anti-MASP1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB10144-100EA 0 EUR
BIRBORB10144-100 BIRBORB10144-200
Anti-MASP1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: MASP1 (mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR