Anti-CASP6 Mouse polyclonal antibody
Toimittaja: Biorbyt
BIRBORB10240-100EA 0 EUR
BIRBORB10240-100 BIRBORB10240-200
Anti-CASP6 Mouse polyclonal antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat