Anti-CTSG Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB10253-100EA 0 EUR
BIRBORB10253-100
Anti-CTSG Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CTSG (cathepsin G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now