Anti-HSPB1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB10845-200EA 0 EUR
BIRBORB10845-200 BIRBORB10845-100
Anti-HSPB1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR