Anti-IL12 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB10893-200EA 0 EUR
BIRBORB10893-200 BIRBORB10893-100
Anti-IL12 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000(based on 0.5 mg/ml)
Type: Primary
Antigen: IL12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat