Anti-IRS4 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB10939-5EA 51 EUR
BIRBORB10939-5 BIRBORB10939-100 BIRBORB10939-200 BIRBORB10939-20
Anti-IRS4 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IRS4 (insulin receptor substrate 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR