Anti-CCL2 Rabbit polyclonal antibody
Toimittaja: Biorbyt
BIRBORB11028-200EA 0 EUR
BIRBORB11028-200 BIRBORB11028-100
Anti-CCL2 Rabbit polyclonal antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat