Anti-MMP9 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11064-20EA 0 EUR
BIRBORB11064-20 BIRBORB11064-5 BIRBORB11064-100 BIRBORB11064-200
Anti-MMP9 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:10000; Western Blot: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:100 (based on 0.5 mg/ml)

Type: Primary
Antigen: MMP9 (matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR