Anti-PPP2R1A Mouse Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11288-200EA 522 EUR
BIRBORB11288-200 BIRBORB11288-100
Anti-PPP2R1A Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR