Anti-RHOA Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11334-100EA 0 EUR
BIRBORB11334-100 BIRBORB11334-200
Anti-RHOA Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: RHOA (ras homolog family member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR