Anti-SPINDLY Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11410-100EA 334 EUR
BIRBORB11410-100 BIRBORB11410-200
Anti-SPINDLY Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SPINDLY
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR