Anti-STAT3 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11428-100EA 306 EUR
BIRBORB11428-100
Anti-STAT3 Rabbit Polyclonal Antibody
Vasta-aineet
  • Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR