Anti-CAV1 Goat Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11600-100EA 0 EUR
BIRBORB11600-100
Anti-CAV1 Goat Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:2000

Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR