Anti-RAB14 Goat Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB11627-100EA 0 EUR
BIRBORB11627-100
Anti-RAB14 Goat Polyclonal Antibody
Vasta-aineet
Western Blot: 1:250-1:2000; Immunofluorescence: 1:25-1:200
Type: Primary
Antigen: RAB14 (RAB14, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now