Anti-TUBA4A Goat Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11635-100EA 0 EUR
BIRBORB11635-100
Anti-TUBA4A Goat Polyclonal Antibody
Vasta-aineet
Western Blot: 1:250-1:2000; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: TUBA4A (tubulin, alpha 4a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR