Anti-GSTT1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB11657-100EA 0 EUR
BIRBORB11657-100
Anti-GSTT1 Rabbit Polyclonal Antibody
Vasta-aineet
0.1-0.5 ?g/ml

Type: Primary
Antigen: GSTT1 (glutathione S-transferase theta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR