Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin))

Toimittaja: Biorbyt
BIRBORB11925-100EA 0 EUR
BIRBORB11925-100 BIRBORB11925-50
Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR