Anti-CD4 Mouse Monoclonal Antibody (Biotin)

Toimittaja: Biorbyt
BIRBORB11934-100EA 0 EUR
BIRBORB11934-100 BIRBORB11934-500
Anti-CD4 Mouse Monoclonal Antibody (Biotin)
Vasta-aineet
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR