Anti-ANPEP Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB11962-500EA 406 EUR
BIRBORB11962-500 BIRBORB11962-100
Anti-ANPEP Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR