Anti-FUT4 Mouse Monoclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB11979-50EA 0 EUR
BIRBORB11979-50 BIRBORB11979-100
Anti-FUT4 Mouse Monoclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR