Anti-FCGR3A Mouse monoclonal antibody

Toimittaja: Biorbyt
BIRBORB11980-500EA 0 EUR
BIRBORB11980-500 BIRBORB11980-100
Anti-FCGR3A Mouse monoclonal antibody
Vasta-aineet
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR