Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: Biorbyt
BIRBORB11991-50EA 0 EUR
BIRBORB11991-50 BIRBORB11991-100
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR