Anti-CD19 Mouse Monoclonal Antibody (APC (Allophycocyanin))

Toimittaja: Biorbyt
BIRBORB11993-100EA 0 EUR
BIRBORB11993-100 BIRBORB11993-50
Anti-CD19 Mouse Monoclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR