Anti-CD19 Mouse Monoclonal Antibody (Biotin)
Toimittaja: Biorbyt
BIRBORB11995-100EA 0 EUR
BIRBORB11995-100 BIRBORB11995-500
Anti-CD19 Mouse Monoclonal Antibody (Biotin)
Vasta-aineet
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human