Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB12027-50EA 0 EUR
BIRBORB12027-50 BIRBORB12027-100
Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR