Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)

Toimittaja: Biorbyt
BIRBORB12054-100EA 0 EUR
BIRBORB12054-100 BIRBORB12054-50
Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®)
Vasta-aineet
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR