Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB12073-100EA 0 EUR
BIRBORB12073-100 BIRBORB12073-50
Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: CD45RO (CD45RO)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR