Anti-ICAM1 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB12088-100EA 0 EUR
BIRBORB12088-100 BIRBORB12088-500
Anti-ICAM1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR