Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: Biorbyt
BIRBORB12096-100EA 0 EUR
BIRBORB12096-100 BIRBORB12096-50
Anti-NCAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR