Anti-B3GAT1 Mouse Monoclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB12098-100EA 0 EUR
BIRBORB12098-100 BIRBORB12098-50
Anti-B3GAT1 Mouse Monoclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR