Anti-CD63 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB12112-500EA 406 EUR
BIRBORB12112-500 BIRBORB12112-100
Anti-CD63 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR