Anti-CD4/CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)

Toimittaja: Biorbyt
BIRBORB12161-20EA 218 EUR
BIRBORB12161-20 BIRBORB12161-50
Anti-CD4/CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Vasta-aineet
Type: Primary
Antigen: CD4/CD45RA
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR