Anti-IgG1 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB12185-500EA 276 EUR
BIRBORB12185-500 BIRBORB12185-100
Anti-IgG1 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IgG1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR