Anti-IgG2a Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB12192-100EA 0 EUR
BIRBORB12192-100 BIRBORB12192-500
Anti-IgG2a Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IgG2a
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR